siRNA Details
Gene Symbol: TXNDC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160047.1 | NP_001153519.1 | 3 | 533 | ||
NM_020784.2 | NP_065835.2 | 3 | 533 | |||
XM_017021505.1 | XP_016876994.1 | 3 | 513 | |||
GenBank mRNA | AB037765.2 | - | 3 | 445 | ||
AK097748.1 | - | 3 | 320 | |||
BC137081.1 | - | 3 | 413 | 9021078 | ||
BC137082.1 | - | 3 | 413 | 9021079 | ||
BC142650.1 | - | 3 | 456 | 40148019 | ||
BC144469.1 | - | 3 | 413 | 9052996 | ||
BC150205.1 | - | 3 | 454 | 40148865 | ||
BC152428.1 | - | 3 | 445 | 8862542 | ||
DA231411.1 | - | 3 | 533 | |||
DB083799.1 | - | 3 | 513 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
