siRNA Details
Gene Symbol: AIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001624.3 | NP_001615.2 | 11, 12 | 4585 | ||
XM_005266839.3 | XP_005266896.1 | 13, 14 | 5423 | |||
XM_017010333.1 | XP_016865822.1 | 11, 12 | 4810 | |||
GenBank mRNA | AK297008.1 | - | 1, 2 | 145 | ||
AK300443.1 | - | 5, 6 | 620 | |||
AK316287.1 | - | 5, 6 | 621 | |||
BC062788.1 | - | 5, 6 | 643 | |||
U83115.1 | - | 11, 12 | 3840 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
