siRNA Details
Gene Symbol: EXOC6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321729.1 | NP_001308658.1 | 18 | 2077 | ||
NM_001321730.1 | NP_001308659.1 | 17 | 1942 | |||
NM_001321731.1 | NP_001308660.1 | 18 | 2077 | |||
NM_001321733.1 | NP_001308662.1 | 17 | 1942 | |||
NM_001321734.1 | NP_001308663.1 | 17 | 1911 | |||
NM_015189.2 | NP_056004.1 | 18 | 2077 | |||
NR_135773.1 | 18 | 2077 | ||||
XM_005264224.1 | XP_005264281.1 | 11 | 1369 | |||
XM_011532711.2 | XP_011531013.1 | 18 | 2073 | |||
XM_011532712.2 | XP_011531014.1 | 18 | 2073 | |||
XM_017003641.1 | XP_016859130.1 | 14 | 1511 | |||
XM_017003642.1 | XP_016859131.1 | 12 | 1124 | |||
GenBank mRNA | AB023136.1 | - | 18 | 2060 | ||
AK023791.1 | - | 6 | 638 | |||
AK026474.1 | - | 15 | 1572 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
