siRNA Details
Gene Symbol: EXOC6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321729.1 | NP_001308658.1 | 14 | 1560 | ||
NM_001321730.1 | NP_001308659.1 | 14 | 1560 | |||
NM_001321731.1 | NP_001308660.1 | 14 | 1560 | |||
NM_001321733.1 | NP_001308662.1 | 14 | 1560 | |||
NM_001321734.1 | NP_001308663.1 | 13 | 1394 | |||
NM_015189.2 | NP_056004.1 | 14 | 1560 | |||
NR_135773.1 | 14 | 1560 | ||||
NR_135774.1 | 14 | 1560 | ||||
XM_005264224.1 | XP_005264281.1 | 7 | 852 | |||
XM_011532711.2 | XP_011531013.1 | 14 | 1556 | |||
XM_011532712.2 | XP_011531014.1 | 14 | 1556 | |||
XM_017003641.1 | XP_016859130.1 | 10 | 994 | |||
XM_017003642.1 | XP_016859131.1 | 8 | 607 | |||
GenBank mRNA | AB023136.1 | - | 14 | 1543 | ||
AK023791.1 | - | 2 | 121 | |||
AK026474.1 | - | 11 | 1055 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
