siRNA Details
Gene Symbol: MYH7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020884.4 | NP_065935.3 | 45 | 6528 | ||
XM_006723840.3 | XP_006723903.1 | 43 | 6187 | |||
XM_011528941.2 | XP_011527243.1 | 43 | 6211 | |||
XM_011528947.2 | XP_011527249.1 | 37 | 6584 | |||
XM_011528948.2 | XP_011527250.1 | 37 | 5743 | |||
XM_017027986.1 | XP_016883475.1 | 44 | 6229 | |||
XM_017027987.1 | XP_016883476.1 | 39 | 5762 | |||
GenBank mRNA | AB040945.2 | - | 43 | 6237 | ||
BC007808.1 | - | 18 | 3215 | |||
BC151242.1 | - | 43 | 6249 | 8860515 | ||
BG189104.1 | - | 4 | 567 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
