siRNA Details
Gene Symbol: MYH7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020884.4 | NP_065935.3 | 43 | 6156 | ||
XM_006723840.3 | XP_006723903.1 | 41 | 5797 | |||
XM_011528941.2 | XP_011527243.1 | 41 | 5821 | |||
XM_011528947.2 | XP_011527249.1 | 35 | 6194 | |||
XM_011528948.2 | XP_011527250.1 | 35 | 5353 | |||
XM_017027986.1 | XP_016883475.1 | 42 | 5857 | |||
XM_017027987.1 | XP_016883476.1 | 37 | 5372 | |||
GenBank mRNA | AB040945.2 | - | 41 | 5865 | ||
BC007808.1 | - | 16 | 2843 | |||
BC151242.1 | - | 41 | 5877 | 8860515 | ||
BG189104.1 | - | 2 | 177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
