siRNA Details
Gene Symbol: UNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080419.2 | NP_001073888.2 | 16 | 3330 | ||
XM_006722145.3 | XP_006722208.1 | 19 | 7675 | |||
XM_006722148.2 | XP_006722211.1 | 17 | 3471 | |||
XM_011525400.1 | XP_011523702.1 | 17 | 3399 | |||
XM_011525401.2 | XP_011523703.1 | 11 | 2487 | |||
XM_017025247.1 | XP_016880736.1 | 19 | 7711 | |||
XM_017025248.1 | XP_016880737.1 | 18 | 7613 | |||
XM_017025249.1 | XP_016880738.1 | 18 | 3756 | |||
XM_017025250.1 | XP_016880739.1 | 17 | 3658 | |||
GenBank mRNA | AB051540.1 | - | 16 | 3313 | ||
BC053362.1 | - | 16 | 3313 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
