siRNA Details
Gene Symbol: ANKRD36C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310154.1 | NP_001297083.1 | 69 | 4621 | ||
GenBank mRNA | AK123626.1 | - | 16 | 1472 | ||
AL832836.1 | - | 29 | 1426 | |||
AL832977.1 | - | 40 | 2002 | |||
U28831.1 | - | 3 | 128 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
