siRNA Details
Gene Symbol: KIF21B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252100.1 | NP_001239029.1 | 35 | 8246 | ||
NM_001252102.1 | NP_001239031.1 | 35 | 7630 | |||
NM_001252103.1 | NP_001239032.1 | 34 | 7591 | |||
NM_017596.3 | NP_060066.2 | 34 | 8207 | |||
GenBank mRNA | AB007918.2 | - | 34 | 8207 | ||
AL137395.1 | - | 1 | 1974 | |||
BC052964.1 | - | 1 | 1659 | |||
BC078676.1 | - | 6 | 3214 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
