siRNA Details
Gene Symbol: CAND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162499.1 | NP_001155971.1 | 15 | 4536 | ||
NM_012298.2 | NP_036430.1 | 13 | 4185 | |||
XM_011533504.2 | XP_011531806.1 | 15 | 4650 | |||
GenBank mRNA | AB014567.1 | - | 13 | 4113 | ||
BX648800.1 | - | 6 | 1689 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
