siRNA Details
Gene Symbol: RUBCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145642.3 | NP_001139114.1 | 19 | 2969 | ||
NM_014687.2 | NP_055502.1 | 18 | 2765 | |||
XM_005269374.3 | XP_005269431.1 | 19 | 2805 | |||
XM_006713827.3 | XP_006713890.1 | 20 | 2881 | |||
XM_006713828.3 | XP_006713891.1 | 19 | 2835 | |||
XM_006713829.3 | XP_006713892.1 | 20 | 3196 | |||
XM_006713830.3 | XP_006713893.1 | 19 | 2729 | |||
XM_006713831.3 | XP_006713894.1 | 19 | 3042 | |||
XM_017007543.1 | XP_016863032.1 | 18 | 2680 | |||
XM_017007544.1 | XP_016863033.1 | 18 | 2651 | |||
XM_017007545.1 | XP_016863034.1 | 18 | 2967 | |||
GenBank mRNA | AK295625.1 | - | 5 | 452 | ||
BC014173.2 | - | 13 | 1921 | |||
D86979.3 | - | 19 | 2658 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
