siRNA Details
Gene Symbol: RUBCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145642.3 | NP_001139114.1 | 9 | 1690 | ||
NM_014687.2 | NP_055502.1 | 8 | 1486 | |||
XM_005269374.3 | XP_005269431.1 | 9 | 1526 | |||
XM_006713827.3 | XP_006713890.1 | 9 | 1527 | |||
XM_006713828.3 | XP_006713891.1 | 8 | 1481 | |||
XM_006713829.3 | XP_006713892.1 | 9 | 1842 | |||
XM_006713830.3 | XP_006713893.1 | 8 | 1375 | |||
XM_006713831.3 | XP_006713894.1 | 8 | 1688 | |||
XM_017007543.1 | XP_016863032.1 | 7 | 1326 | |||
XM_017007544.1 | XP_016863033.1 | 8 | 1372 | |||
XM_017007545.1 | XP_016863034.1 | 8 | 1688 | |||
GenBank mRNA | BC014173.2 | - | 2 | 567 | ||
BX647598.1 | - | 8 | 1263 | |||
D86979.3 | - | 9 | 1379 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
