siRNA Details
Gene Symbol: KIAA1147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080392.1 | NP_001073861.1 | 6 | 938 | ||
GenBank mRNA | AB032973.2 | - | 6 | 927 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
