siRNA Details
Gene Symbol: FAM199X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207318.3 | NP_997201.1 | 6 | 2564 | ||
XM_005262079.2 | XP_005262136.1 | 6 | 2432 | |||
XM_017029270.1 | XP_016884759.1 | 6 | 2133 | |||
GenBank mRNA | AK091221.1 | - | 7 | 2411 | ||
BC016683.1 | - | 4 | 1655 | |||
BC017665.1 | - | 6 | 2419 | 3903800 | ||
BC035886.1 | - | 5 | 1955 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
