siRNA Details
Gene Symbol: DIP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173602.2 | NP_775873.2 | 23 | 2903 | ||
XM_005269044.1 | XP_005269101.1 | 23 | 2906 | |||
XM_006719520.3 | XP_006719583.1 | 24 | 2879 | |||
XM_006719521.3 | XP_006719584.1 | 23 | 2716 | |||
XM_011538594.2 | XP_011536896.1 | 24 | 2932 | |||
XM_011538595.2 | XP_011536897.1 | 22 | 2764 | |||
XM_011538596.2 | XP_011536898.1 | 23 | 2719 | |||
XM_017019699.1 | XP_016875188.1 | 24 | 2914 | |||
XM_017019700.1 | XP_016875189.1 | 23 | 2888 | |||
XM_017019701.1 | XP_016875190.1 | 23 | 2885 | |||
XM_017019702.1 | XP_016875191.1 | 21 | 2735 | |||
XM_017019703.1 | XP_016875192.1 | 22 | 2682 | |||
XM_017019704.1 | XP_016875193.1 | 22 | 2690 | |||
XM_017019705.1 | XP_016875194.1 | 22 | 2672 | |||
GenBank mRNA | AB040896.2 | - | 14 | 1517 | ||
AK097369.1 | - | 5 | 459 | |||
AK126758.1 | - | 12 | 1274 | |||
BC007671.2 | - | 7 | 827 | |||
BC030156.2 | - | 7 | 827 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
