siRNA Details
Gene Symbol: COQ5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032314.3 | NP_115690.3 | 7 | 1037 | ||
XM_006719639.2 | XP_006719702.1 | 8 | 1137 | |||
GenBank mRNA | AK057777.1 | - | 4 | 539 | ||
AK222610.1 | - | 7 | 1027 | |||
AK225242.1 | - | 7 | 1027 | |||
AK293656.1 | - | 7 | 1058 | |||
AK298214.1 | - | 6 | 815 | |||
BC004916.2 | - | 7 | 1013 | |||
BC107874.1 | - | 7 | 1031 | 6452052 | ||
BX647562.1 | - | 7 | 1026 | |||
EU700459.1 | - | 7 | 1031 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
