siRNA Details
Gene Symbol: SVEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153366.3 | NP_699197.3 | 9 | 2200 | ||
GenBank mRNA | AJ619977.1 | - | 9 | 2026 | ||
AK122605.1 | - | 9 | 2196 | |||
AY243503.1 | - | 9 | 2026 | |||
BC030816.1 | - | 9 | 2200 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
