siRNA Details
Gene Symbol: SVEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153366.3 | NP_699197.3 | 15 | 3041 | ||
GenBank mRNA | AJ619977.1 | - | 15 | 2867 | ||
AY243503.1 | - | 15 | 2867 | |||
CX165144.1 | - | 1 | 95 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
