siRNA Details
Gene Symbol: SCN1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165963.1 | NP_001159435.1 | 26 | 6251 | ||
NM_001165964.1 | NP_001159436.1 | 26 | 6167 | |||
NM_001202435.1 | NP_001189364.1 | 28 | 6460 | |||
NM_006920.4 | NP_008851.3 | 26 | 6218 | |||
XM_011511602.2 | XP_011509904.1 | 27 | 6527 | |||
XM_011511605.2 | XP_011509907.1 | 28 | 6591 | |||
XM_011511606.2 | XP_011509908.1 | 28 | 6535 | |||
XM_017004644.1 | XP_016860133.1 | 29 | 6706 | |||
XM_017004650.1 | XP_016860139.1 | 27 | 6493 | |||
XM_017004651.1 | XP_016860140.1 | 28 | 6398 | |||
XM_017004652.1 | XP_016860141.1 | 27 | 6438 | |||
XM_017004653.1 | XP_016860142.1 | 28 | 6545 | |||
XM_017004654.1 | XP_016860143.1 | 28 | 6626 | |||
GenBank mRNA | AK293759.1 | - | 2 | 1509 | ||
AK294900.1 | - | 6 | 1994 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
