siRNA Details
Gene Symbol: PHF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014660.3 | NP_055475.2 | 17 | 4074 | ||
NR_033435.1 | 16 | 3286 | ||||
GenBank mRNA | AB018326.1 | - | 17 | 4029 | ||
BC152414.1 | - | 17 | 4029 | 8862519 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
