siRNA Details
Gene Symbol: PHF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014660.3 | NP_055475.2 | 10 | 2337 | ||
NR_033435.1 | 9 | 1549 | ||||
NR_033436.1 | 9 | 1549 | ||||
XM_017012811.1 | XP_016868300.1 | 10 | 2375 | |||
XM_017012812.1 | XP_016868301.1 | 10 | 2375 | |||
XM_017012813.1 | XP_016868302.1 | 10 | 2374 | |||
XM_017012814.1 | XP_016868303.1 | 10 | 2374 | |||
XM_017012815.1 | XP_016868304.1 | 10 | 2374 | |||
XM_017012816.1 | XP_016868305.1 | 10 | 2373 | |||
XM_017012817.1 | XP_016868306.1 | 9 | 1584 | |||
GenBank mRNA | AB018326.1 | - | 10 | 2292 | ||
AK294419.1 | - | 9 | 1549 | |||
AK295461.1 | - | 9 | 1532 | |||
BC152414.1 | - | 10 | 2292 | 8862519 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001168382, NM_029404, AK003209, AK173024, AK016517, and others.
Gene Ontology Categories:
