siRNA Details
Gene Symbol: TTBK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032538.1 | NP_115927.1 | 13 | 1772 | ||
XM_011514947.2 | XP_011513249.1 | 12 | 1586 | |||
XM_017011364.1 | XP_016866853.1 | 11 | 1823 | |||
XM_017011365.1 | XP_016866854.1 | 11 | 1823 | |||
XM_017011366.1 | XP_016866855.1 | 12 | 1562 | |||
XM_017011367.1 | XP_016866856.1 | 11 | 1823 | |||
GenBank mRNA | AB058758.1 | - | 11 | 1341 | ||
AB218664.1 | - | 14 | 1576 | |||
AK131217.1 | - | 12 | 1502 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
