siRNA Details
Gene Symbol: SH3D19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009555.3 | NP_001009555.3 | 11 | 2254 | ||
NM_001128923.1 | NP_001122395.1 | 11 | 2242 | |||
NM_001128924.1 | NP_001122396.1 | 4 | 1245 | |||
NM_001243349.1 | NP_001230278.1 | 11 | 2163 | |||
XM_005262767.2 | XP_005262824.1 | 12 | 2115 | |||
XM_011531646.1 | XP_011529948.1 | 10 | 1901 | |||
XM_011531647.1 | XP_011529949.1 | 9 | 1793 | |||
XM_011531648.1 | XP_011529950.1 | 12 | 2228 | |||
XM_011531649.1 | XP_011529951.1 | 9 | 1793 | |||
XM_017007778.1 | XP_016863267.1 | 12 | 2219 | |||
XM_017007779.1 | XP_016863268.1 | 9 | 1784 | |||
XM_017007780.1 | XP_016863269.1 | 12 | 2219 | |||
XM_017007781.1 | XP_016863270.1 | 11 | 2145 | |||
XM_017007782.1 | XP_016863271.1 | 11 | 2145 | |||
XM_017007783.1 | XP_016863272.1 | 12 | 2115 | |||
GenBank mRNA | AK092528.1 | - | 4 | 475 | ||
AK128493.1 | - | 4 | 1094 | |||
AK291025.1 | - | 3 | 342 | |||
AK294476.1 | - | 11 | 2163 | |||
AL133047.1 | - | 1 | 18 | |||
BC010110.2 | - | 4 | 516 | |||
BC085613.1 | - | 2 | 233 | |||
BC108890.1 | - | 5 | 1353 | 40001764 | ||
BC108891.1 | - | 5 | 1353 | 40001765 | ||
BC108892.1 | - | 5 | 1353 | 40001766 | ||
BC108893.1 | - | 4 | 1245 | 40001767 | ||
BX647422.1 | - | 11 | 2166 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
