siRNA Details
Gene Symbol: PHACTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100164.1 | NP_001093634.1 | 12 | 2109 | ||
NM_001100165.1 | NP_001093635.1 | 11 | 1869 | |||
NM_001100166.1 | NP_001093636.1 | 11 | 1767 | |||
NM_014721.2 | NP_055536.2 | 12 | 2007 | |||
GenBank mRNA | AB014580.1 | - | 12 | 1924 | ||
AK297799.1 | - | 12 | 1914 | |||
AK298263.1 | - | 12 | 2109 | |||
AK300221.1 | - | 11 | 1762 | |||
BC150630.1 | - | 12 | 1914 | 9057000 | ||
BC152416.1 | - | 12 | 1924 | 8862521 | ||
CR749345.1 | - | 11 | 1807 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001195066, NM_001195096, NM_001195065, NM_001033257, BC130264, and others.
Gene Ontology Categories:
