siRNA Details
Gene Symbol: IBTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300906.1 | NP_001287835.1 | 10 | 1860 | ||
NM_015525.3 | NP_056340.2 | 10 | 1860 | |||
XM_006715453.2 | XP_006715516.1 | 10 | 2400 | |||
XM_006715454.2 | XP_006715517.1 | 10 | 2400 | |||
GenBank mRNA | AB037838.2 | - | 9 | 1584 | ||
BC092410.1 | - | 9 | 1460 | |||
BC113696.1 | - | 9 | 1389 | 8322748 | ||
BC113698.1 | - | 9 | 1389 | 8322750 | ||
BC143736.1 | - | 9 | 1389 | 9052250 | ||
BC143745.1 | - | 9 | 1390 | 9052259 | ||
BC143748.1 | - | 9 | 1389 | |||
DQ005633.1 | - | 10 | 1860 | |||
DQ005634.1 | - | 10 | 1860 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
