siRNA Details
Gene Symbol: ZCCHC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185059.1 | NP_001171988.1 | 26 | 4447 | ||
NM_001185074.1 | NP_001172003.1 | 19 | 3849 | |||
NM_024617.3 | NP_078893.2 | 26 | 4557 | |||
XM_005252207.3 | XP_005252264.1 | 26 | 4509 | |||
XM_006717283.1 | XP_006717346.1 | 26 | 4509 | |||
XM_011519011.2 | XP_011517313.1 | 26 | 4509 | |||
XM_011519012.2 | XP_011517314.1 | 26 | 4457 | |||
XM_011519013.2 | XP_011517315.1 | 26 | 5126 | |||
XM_011519014.2 | XP_011517316.1 | 26 | 4728 | |||
XM_011519015.1 | XP_011517317.1 | 26 | 4509 | |||
XM_011519016.1 | XP_011517318.1 | 26 | 4509 | |||
XM_017015130.1 | XP_016870619.1 | 26 | 5125 | |||
XM_017015131.1 | XP_016870620.1 | 26 | 4767 | |||
XM_017015132.1 | XP_016870621.1 | 27 | 4672 | |||
GenBank mRNA | AB051498.1 | - | 18 | 3095 | ||
AK023471.1 | - | 11 | 1143 | |||
AL832026.1 | - | 19 | 3813 | |||
AL832193.1 | - | 27 | 4599 | |||
AL832965.1 | - | 2 | 310 | |||
AL832988.1 | - | 13 | 1392 | |||
BC144523.1 | - | 24 | 4241 | |||
BX641077.1 | - | 25 | 5907 | |||
CR933643.1 | - | 26 | 4421 | |||
CR933644.1 | - | 26 | 4999 | |||
CR936608.1 | - | 26 | 4501 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
