siRNA Details
Gene Symbol: ZCCHC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185059.1 | NP_001171988.1 | 22 | 3842 | ||
NM_001185074.1 | NP_001172003.1 | 15 | 3244 | |||
NM_024617.3 | NP_078893.2 | 22 | 3952 | |||
XM_005252207.3 | XP_005252264.1 | 22 | 3904 | |||
XM_005252208.2 | XP_005252265.1 | 22 | 3904 | |||
XM_006717283.1 | XP_006717346.1 | 22 | 3904 | |||
XM_011519011.2 | XP_011517313.1 | 22 | 3904 | |||
XM_011519012.2 | XP_011517314.1 | 22 | 3852 | |||
XM_011519013.2 | XP_011517315.1 | 22 | 4521 | |||
XM_011519014.2 | XP_011517316.1 | 22 | 4123 | |||
XM_011519015.1 | XP_011517317.1 | 22 | 3904 | |||
XM_011519016.1 | XP_011517318.1 | 22 | 3904 | |||
XM_017015130.1 | XP_016870619.1 | 22 | 4520 | |||
XM_017015131.1 | XP_016870620.1 | 22 | 4162 | |||
XM_017015132.1 | XP_016870621.1 | 23 | 4067 | |||
GenBank mRNA | AB051498.1 | - | 14 | 2490 | ||
AK023471.1 | - | 7 | 538 | |||
AK055948.1 | - | 10 | 1802 | |||
AL832026.1 | - | 15 | 3208 | |||
AL832193.1 | - | 23 | 3994 | |||
AL832988.1 | - | 9 | 787 | |||
BC144523.1 | - | 20 | 3636 | |||
BX641077.1 | - | 21 | 5302 | |||
CR933643.1 | - | 22 | 3816 | |||
CR933644.1 | - | 22 | 4394 | |||
CR936608.1 | - | 22 | 3896 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
