siRNA Details
Gene Symbol: ARID5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244638.1 | NP_001231567.1 | 7 | 1948 | ||
NM_032199.2 | NP_115575.1 | 10 | 2864 | |||
XM_011540262.1 | XP_011538564.1 | 9 | 2633 | |||
XM_017016765.1 | XP_016872254.1 | 5 | 1622 | |||
GenBank mRNA | BC036831.1 | - | 1 | 992 | ||
BX537690.1 | - | 1 | 371 | |||
BX641020.1 | - | 1 | 652 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
