siRNA Details
Gene Symbol: YLPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019589.2 | NP_062535.2 | 10, 11 | 5479 | ||
XM_005267860.3 | XP_005267917.1 | 10, 11 | 5523 | |||
XM_011536966.2 | XP_011535268.1 | 11, 12 | 4638 | |||
XM_011536967.1 | XP_011535269.1 | 9, 10 | 3405 | |||
GenBank mRNA | AK090435.1 | - | 8, 9 | 4235 | ||
AK297582.1 | - | 6, 7 | 1050 | |||
AL832365.1 | - | 7, 8 | 4014 | |||
BC007792.1 | - | 1, 2 | 76 | |||
L40403.1 | - | 4, 5 | 582 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
