siRNA Details
Gene Symbol: YLPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019589.2 | NP_062535.2 | 11, 12 | 5619 | ||
XM_005267860.3 | XP_005267917.1 | 11, 12 | 5663 | |||
XM_011536966.2 | XP_011535268.1 | 12, 13 | 4778 | |||
XM_011536967.1 | XP_011535269.1 | 10, 11 | 3545 | |||
GenBank mRNA | AK090435.1 | - | 9, 10 | 4375 | ||
AK297582.1 | - | 7, 8 | 1190 | |||
AL832365.1 | - | 8, 9 | 4154 | |||
BC007792.1 | - | 2, 3 | 216 | |||
L40403.1 | - | 5, 6 | 722 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
