siRNA Details
Gene Symbol: LOC105371998 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU620199.1 | - | 1 | 216 |
Gene Symbol: SPIRE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128626.1 | NP_001122098.1 | 17 | 4356 | ||
NM_001128627.1 | NP_001122099.1 | 16 | 4012 | |||
NM_020148.2 | NP_064533.3 | 16 | 4314 | |||
XM_005258122.4 | XP_005258179.1 | 17 | 4671 | |||
XM_011525701.1 | XP_011524003.1 | 18 | 4719 | |||
XM_011525702.1 | XP_011524004.1 | 18 | 4231 | |||
GenBank mRNA | AB032961.2 | - | 17 | 4409 | ||
AJ277587.1 | - | 14 | 3901 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
