siRNA Details
Gene Symbol: CC2D2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080522.2 | NP_001073991.2 | 23 | 3095 | ||
XM_005248177.1 | XP_005248234.1 | 22 | 3050 | |||
XM_011513872.2 | XP_011512174.1 | 22 | 3050 | |||
XM_017008482.1 | XP_016863971.1 | 20 | 2844 | |||
GenBank mRNA | AB037766.1 | - | 20 | 2833 | ||
AK023876.1 | - | 14 | 2005 | |||
AK025543.1 | - | 8 | 1180 | |||
BX647334.1 | - | 22 | 3020 | |||
EU450799.1 | - | 23 | 2922 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
