siRNA Details
Gene Symbol: CC2D2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080522.2 | NP_001073991.2 | 11 | 1148 | ||
XM_005248177.1 | XP_005248234.1 | 10 | 1103 | |||
XM_011513872.2 | XP_011512174.1 | 10 | 1103 | |||
XM_017008482.1 | XP_016863971.1 | 8 | 897 | |||
GenBank mRNA | AB037766.1 | - | 8 | 886 | ||
AK023876.1 | - | 2 | 58 | |||
AK096047.1 | - | 10 | 1087 | |||
AK309178.1 | - | 10 | 1078 | |||
BC103710.1 | - | 8 | 876 | |||
BX647334.1 | - | 10 | 1083 | |||
EU450799.1 | - | 11 | 975 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
