siRNA Details
Gene Symbol: PRR5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160167.1 | NP_001153639.1 | 9 | 1880 | ||
NM_001160168.1 | NP_001153640.1 | 6 | 1279 | |||
NM_001160169.1 | NP_001153641.1 | 7 | 1489 | |||
NM_024841.4 | NP_079117.3 | 10 | 1866 | |||
GenBank mRNA | AK024275.1 | - | 6 | 1226 | ||
AK055042.1 | - | 9 | 1767 | |||
BC008922.2 | - | 2 | 937 | |||
BC125122.1 | - | 1 | 809 | |||
BX640959.1 | - | 10 | 1847 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
