siRNA Details
Gene Symbol: ATXN7L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020218.1 | NP_064603.1 | 5, 6 | 571 | ||
XM_011525038.2 | XP_011523340.1 | 5, 6 | 1165 | |||
XM_011525044.2 | XP_011523346.1 | 5, 6 | 1165 | |||
XM_017024880.1 | XP_016880369.1 | 6, 7 | 743 | |||
XM_017024881.1 | XP_016880370.1 | 6, 7 | 1042 | |||
XM_017024883.1 | XP_016880372.1 | 6, 7 | 746 | |||
XM_017024885.1 | XP_016880374.1 | 6, 7 | 746 | |||
GenBank mRNA | BC037418.1 | - | 5, 6 | 478 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
