siRNA Details
Gene Symbol: ATXN7L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098833.1 | NP_001092303.1 | 10 | 805 | ||
NM_020218.1 | NP_064603.1 | 10 | 826 | |||
XM_011525038.2 | XP_011523340.1 | 10 | 1477 | |||
XM_011525044.2 | XP_011523346.1 | 10 | 1420 | |||
XM_011525046.2 | XP_011523348.1 | 10 | 1399 | |||
XM_017024880.1 | XP_016880369.1 | 11 | 1055 | |||
XM_017024881.1 | XP_016880370.1 | 11 | 1354 | |||
XM_017024882.1 | XP_016880371.1 | 10 | 1456 | |||
XM_017024883.1 | XP_016880372.1 | 11 | 1058 | |||
XM_017024884.1 | XP_016880373.1 | 10 | 1080 | |||
XM_017024885.1 | XP_016880374.1 | 11 | 1001 | |||
XM_017024886.1 | XP_016880375.1 | 11 | 980 | |||
GenBank mRNA | AK056002.1 | - | 7 | 349 | ||
AK126117.1 | - | 10 | 1023 | |||
AK296689.1 | - | 4 | 238 | |||
AL390158.1 | - | 5 | 235 | |||
BC024695.2 | - | 3 | 163 | |||
BC037418.1 | - | 10 | 733 | |||
BC113595.1 | - | 10 | 805 | 8322647 | ||
BC126113.1 | - | 10 | 805 | 8991829 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
