siRNA Details
Gene Symbol: ATXN7L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098833.1 | NP_001092303.1 | 12 | 2559 | ||
NM_020218.1 | NP_064603.1 | 12 | 2580 | |||
XM_011525038.2 | XP_011523340.1 | 12 | 3231 | |||
XM_011525044.2 | XP_011523346.1 | 12 | 3174 | |||
XM_011525046.2 | XP_011523348.1 | 12 | 3153 | |||
XM_017024880.1 | XP_016880369.1 | 13 | 2809 | |||
XM_017024881.1 | XP_016880370.1 | 13 | 3108 | |||
XM_017024882.1 | XP_016880371.1 | 12 | 3210 | |||
XM_017024883.1 | XP_016880372.1 | 13 | 2812 | |||
XM_017024884.1 | XP_016880373.1 | 12 | 2834 | |||
XM_017024885.1 | XP_016880374.1 | 13 | 2755 | |||
XM_017024886.1 | XP_016880375.1 | 13 | 2734 | |||
GenBank mRNA | AK056002.1 | - | 9 | 2103 | ||
AK074344.1 | - | 1 | 1168 | |||
AL390158.1 | - | 7 | 1988 | |||
BC024695.2 | - | 5 | 1917 | |||
BC037418.1 | - | 12 | 2487 | |||
BC100295.1 | - | 1 | 338 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
