siRNA Details
Gene Symbol: PGGHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025092.4 | NP_079368.3 | 9 | 1544 | ||
XM_011520383.2 | XP_011518685.1 | 12 | 2470 | |||
XM_011520384.2 | XP_011518686.1 | 11 | 2581 | |||
XM_011520385.2 | XP_011518687.1 | 11 | 2584 | |||
XM_017018355.1 | XP_016873844.1 | 10 | 1922 | |||
GenBank mRNA | AK026288.1 | - | 6 | 955 | ||
AK074155.1 | - | 6 | 2706 | |||
AK090428.1 | - | 4 | 1070 | |||
AK296137.1 | - | 5 | 423 | |||
AL832932.1 | - | 2 | 202 | |||
BC109257.2 | - | 6 | 943 | 40024867 | ||
BC109258.2 | - | 6 | 943 | 40024874 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
