siRNA Details
Gene Symbol: PGGHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025092.4 | NP_079368.3 | 10 | 1686 | ||
XM_011520383.2 | XP_011518685.1 | 13 | 2612 | |||
XM_011520384.2 | XP_011518686.1 | 12 | 2723 | |||
XM_017018355.1 | XP_016873844.1 | 11 | 2064 | |||
GenBank mRNA | AK074155.1 | - | 7 | 2848 | ||
AK090428.1 | - | 4 | 1303 | |||
AK296137.1 | - | 6 | 565 | |||
AL832932.1 | - | 3 | 344 | |||
BC048336.1 | - | 1 | 226 | |||
BC066641.1 | - | 9 | 1393 | |||
BC109257.2 | - | 7 | 1085 | 40024867 | ||
BC109258.2 | - | 7 | 1085 | 40024874 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
