siRNA Details
Gene Symbol: PHF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009936.2 | NP_001009936.1 | 3 | 497 | ||
NM_001286840.1 | NP_001273769.1 | 3 | 368 | |||
NM_001286843.1 | NP_001273772.1 | 3 | 497 | |||
NM_015651.2 | NP_056466.1 | 3 | 497 | |||
XM_005251906.2 | XP_005251963.1 | 3 | 305 | |||
XM_011518509.2 | XP_011516811.1 | 3 | 348 | |||
XM_011518511.2 | XP_011516813.1 | 3 | 831 | |||
XM_011518515.2 | XP_011516817.1 | 3 | 832 | |||
XM_011518516.2 | XP_011516818.1 | 3 | 834 | |||
XM_017014612.1 | XP_016870101.1 | 3 | 337 | |||
XM_017014613.1 | XP_016870102.1 | 3 | 835 | |||
GenBank mRNA | AK122744.1 | - | 3 | 357 | ||
BC022374.1 | - | 3 | 357 | |||
BC108663.1 | - | 3 | 464 | |||
BC125076.1 | - | 2 | 269 | 40117657 | ||
BC125077.1 | - | 2 | 269 | 40117658 | ||
BX640713.1 | - | 3 | 369 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
