siRNA Details
Gene Symbol: PHF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009936.2 | NP_001009936.1 | 2 | 413 | ||
NM_001286840.1 | NP_001273769.1 | 2 | 284 | |||
NM_001286843.1 | NP_001273772.1 | 2 | 413 | |||
NM_015651.2 | NP_056466.1 | 2 | 413 | |||
XM_005251906.2 | XP_005251963.1 | 2 | 221 | |||
XM_011518509.2 | XP_011516811.1 | 2 | 264 | |||
XM_011518511.2 | XP_011516813.1 | 2 | 747 | |||
XM_011518515.2 | XP_011516817.1 | 2 | 748 | |||
XM_011518516.2 | XP_011516818.1 | 2 | 750 | |||
XM_017014612.1 | XP_016870101.1 | 2 | 253 | |||
XM_017014613.1 | XP_016870102.1 | 2 | 751 | |||
GenBank mRNA | AK122744.1 | - | 2 | 273 | ||
BC022374.1 | - | 2 | 273 | |||
BC108663.1 | - | 2 | 380 | |||
BC125076.1 | - | 1 | 185 | 40117657 | ||
BC125077.1 | - | 1 | 185 | 40117658 | ||
BM784783.1 | - | 2 | 413 | |||
BX640713.1 | - | 2 | 285 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
