siRNA Details
Gene Symbol: CFAP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008723.1 | NP_001008723.1 | 8 | 1292 | ||
XM_011539377.2 | XP_011537679.1 | 8 | 2011 | |||
XM_017015787.1 | XP_016871276.1 | 8 | 1663 | |||
XM_017015788.1 | XP_016871277.1 | 8 | 2194 | |||
XM_017015789.1 | XP_016871278.1 | 8 | 1281 | |||
XM_017015790.1 | XP_016871279.1 | 8 | 1281 | |||
GenBank mRNA | AK093227.1 | - | 8 | 1280 | ||
AK296463.1 | - | 8 | 1285 | |||
BC036225.1 | - | 8 | 1294 | 5272786 | ||
BC109126.2 | - | 8 | 1246 | 40036085 | ||
BX641120.1 | - | 8 | 1293 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
