siRNA Details
Gene Symbol: GLTSCR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318819.1 | NP_001305748.1 | 6, 7 | 596 | ||
NM_015349.2 | NP_056164.1 | 4, 5 | 329 | |||
GenBank mRNA | AK091434.1 | - | 4, 5 | 221 | ||
AK093526.1 | - | 4, 5 | 940 | |||
AK294706.1 | - | 4, 5 | 221 | |||
AL833540.1 | - | 4, 5 | 329 | |||
BC130302.1 | - | 4, 5 | 252 | 40146333 | ||
CR627378.1 | - | 5, 6 | 294 | |||
D87077.2 | - | 7, 8 | 493 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
