siRNA Details
Gene Symbol: GLTSCR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318819.1 | NP_001305748.1 | 9 | 2410 | ||
NM_015349.2 | NP_056164.1 | 7 | 2143 | |||
GenBank mRNA | AK294706.1 | - | 7 | 2035 | ||
AL833540.1 | - | 7 | 2143 | |||
BC130302.1 | - | 7 | 2066 | 40146333 | ||
CR627378.1 | - | 8 | 2108 | |||
D87077.2 | - | 10 | 2307 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
