siRNA Details
Gene Symbol: TENM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080428.2 | NP_001073897.2 | 22 | 4973 | ||
NM_001122679.1 | NP_001116151.1 | 26 | 5545 | |||
XM_005265950.2 | XP_005266007.1 | 28 | 6192 | |||
XM_005265952.2 | XP_005266009.1 | 24 | 4919 | |||
XM_006714897.2 | XP_006714960.1 | 27 | 6171 | |||
XM_011534604.2 | XP_011532906.1 | 27 | 6006 | |||
XM_017009660.1 | XP_016865149.1 | 28 | 6219 | |||
XM_017009661.1 | XP_016865150.1 | 27 | 6198 | |||
XM_017009662.1 | XP_016865151.1 | 27 | 6009 | |||
XM_017009663.1 | XP_016865152.1 | 25 | 5414 | |||
XM_017009664.1 | XP_016865153.1 | 25 | 5327 | |||
XM_017009665.1 | XP_016865154.1 | 25 | 5123 | |||
XM_017009666.1 | XP_016865155.1 | 24 | 5102 | |||
XM_017009667.1 | XP_016865156.1 | 25 | 5096 | |||
XM_017009668.1 | XP_016865157.1 | 25 | 6633 | |||
XM_017009669.1 | XP_016865158.1 | 25 | 6465 | |||
GenBank mRNA | AB032953.2 | - | 17 | 3682 | ||
BX648178.1 | - | 21 | 4567 | |||
CR936703.1 | - | 22 | 4906 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
