siRNA Details
Gene Symbol: TENM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080428.2 | NP_001073897.2 | 22 | 5207 | ||
NM_001122679.1 | NP_001116151.1 | 26 | 5779 | |||
XM_005265950.2 | XP_005266007.1 | 28 | 6426 | |||
XM_005265952.2 | XP_005266009.1 | 24 | 5153 | |||
XM_006714897.2 | XP_006714960.1 | 27 | 6405 | |||
XM_011534604.2 | XP_011532906.1 | 27 | 6240 | |||
XM_017009660.1 | XP_016865149.1 | 28 | 6453 | |||
XM_017009661.1 | XP_016865150.1 | 27 | 6432 | |||
XM_017009662.1 | XP_016865151.1 | 27 | 6243 | |||
XM_017009663.1 | XP_016865152.1 | 25 | 5648 | |||
XM_017009664.1 | XP_016865153.1 | 25 | 5561 | |||
XM_017009665.1 | XP_016865154.1 | 25 | 5357 | |||
XM_017009666.1 | XP_016865155.1 | 24 | 5336 | |||
XM_017009667.1 | XP_016865156.1 | 25 | 5330 | |||
XM_017009668.1 | XP_016865157.1 | 25 | 6867 | |||
XM_017009669.1 | XP_016865158.1 | 25 | 6699 | |||
GenBank mRNA | AB032953.2 | - | 17 | 3916 | ||
AL137500.1 | - | 1 | 32 | |||
BX648178.1 | - | 21 | 4801 | |||
CR936703.1 | - | 22 | 5140 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
