siRNA Details
Gene Symbol: NOP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291978.1 | NP_001278907.1 | 10, 11 | 1685 | ||
NM_001291979.1 | NP_001278908.1 | 10, 11 | 1685 | |||
NM_003703.2 | NP_003694.1 | 10, 11 | 1685 | |||
GenBank mRNA | AB000467.1 | - | 8, 9 | 1081 | ||
AF040965.1 | - | 10, 11 | 1547 | |||
AK025692.1 | - | 4, 5 | 550 | |||
AK292939.1 | - | 10, 11 | 1685 | |||
BC009760.2 | - | 6, 7 | 879 | |||
BC026035.1 | - | 10, 11 | 1512 | 4878317 | ||
BC053635.1 | - | 10, 11 | 1564 | 5114280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
