siRNA Details
Gene Symbol: NOP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291978.1 | NP_001278907.1 | 17, 18 | 2667 | ||
NM_003703.2 | NP_003694.1 | 17, 18 | 2667 | |||
GenBank mRNA | AB000467.1 | - | 15, 16 | 2063 | ||
AF040965.1 | - | 17, 18 | 2529 | |||
AK024755.1 | - | 7, 8 | 976 | |||
AK025692.1 | - | 11, 12 | 1532 | |||
AK292939.1 | - | 17, 18 | 2667 | |||
BC053635.1 | - | 17, 18 | 2546 | 5114280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
