siRNA Details
Gene Symbol: ARHGAP28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010000.2 | NP_001010000.1 | 12 | 1487 | ||
XM_005258144.1 | XP_005258201.2 | 13 | 1809 | |||
XM_005258145.1 | XP_005258202.2 | 13 | 1809 | |||
XM_005258146.2 | XP_005258203.1 | 12 | 1731 | |||
GenBank mRNA | AB037735.1 | - | 13 | 4760 | ||
AK001174.1 | - | 7 | 816 | |||
AK289791.1 | - | 6 | 747 | |||
AK302026.1 | - | 12 | 1396 | |||
AK308259.1 | - | 12 | 1436 | |||
BC033668.1 | - | 6 | 706 | |||
BC065274.1 | - | 11 | 1248 | 6164396 | ||
BX648684.1 | - | 12 | 1445 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
