siRNA Details
Gene Symbol: TMEM181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_020823.1 | NP_065874.1 | 5 | 752 | ||
XM_005267074.3 | XP_005267131.1 | 5 | 469 | |||
XM_011535999.1 | XP_011534301.1 | 5 | 756 | |||
XM_011536000.1 | XP_011534302.1 | 5 | 756 | |||
XM_011536001.2 | XP_011534303.1 | 5 | 936 | |||
XM_011536002.2 | XP_011534304.1 | 5 | 934 | |||
XM_011536003.2 | XP_011534305.1 | 5 | 756 | |||
GenBank mRNA | AB037844.1 | - | 5 | 754 | ||
AY927471.1 | - | LIT3489 AY927471 |
5 | 869 | ||
AY927472.1 | - | 6 | 1405 | |||
BC033450.1 | - | 4 | 475 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
